Assay Details
| Gene Symbol: CLPP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018050927.1 | XP_017906416.1 | 2 - 3 | 349 | 68 | |
| GenBank mRNA | KA325031.1 | - | 4 - 5 | 863 | 68 | |
More Information
Set Membership: |
|

CLPP
Genomic Map