Assay Details
| Gene Symbol: CYLD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013971026.2 | XP_013826480.1 | 4 - 5 | 1642 | 71 | |
| XM_018061934.1 | XP_017917423.1 | 3 - 4 | 1033 | 71 | ||
| XM_018061935.1 | XP_017917424.1 | 3 - 4 | 1035 | 71 | ||
| XM_018061936.1 | XP_017917425.1 | 3 - 4 | 1537 | 71 | ||
| XM_018061937.1 | XP_017917426.1 | 3 - 4 | 1033 | 71 | ||
| XM_018061938.1 | XP_017917427.1 | 2 - 3 | 347 | 71 | ||
More Information
Set Membership: |
|

CYLD
Genomic Map