Assay Details
| Gene Symbol: NSRP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005693285.3 | XP_005693342.2 | 3 - 4 | 251 | 62 | |
| XM_005693287.3 | XP_005693344.2 | 2 - 3 | 123 | 62 | ||
| GenBank mRNA | JT746828.1 | - | 4 - 5 | 327 | 62 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005693287.3 transcripts.
Set Membership: |
|

NSRP1
Genomic Map