Assay Details
| Gene Symbol: UBE2D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018050305.1 | XP_017905794.1 | 5 - 6 | 924 | 101 | |
| GenBank mRNA | JT714648.1 | - | 6 - 7 | 958 | 101 | |
More Information
Set Membership: |
|

UBE2D2
Genomic Map