Assay Details
Gene Symbol: RAB15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018054064.1 | XP_017909553.1 | 6 - 7 | 635 | 73 | |
XM_018054065.1 | XP_017909554.1 | 5 - 6 | 955 | 73 | ||
GenBank mRNA | KA315467.1 | - | 1 - 2 | 2626 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
