Assay Details
| Gene Symbol: TUBB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013973992.2 | XP_013829446.2 | 2 - 3 | 360 | 75 | |
| GenBank mRNA | JO580471.1 | - | 2 - 3 | 455 | 75 | |
More Information
Set Membership: |
|

TUBB
Genomic Map