Assay Details
| Gene Symbol: ENOSF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013974282.2 | XP_013829736.2 | 7 - 8 | 705 | 57 | |
| XM_018039649.1 | XP_017895138.1 | 5 - 6 | 288 | 57 | ||
| GenBank mRNA | JO428013.1 | - | 9 - 10 | 1966 | 57 | |
More Information
Set Membership: |
|

ENOSF1
Genomic Map