Assay Details
| Gene Symbol: GSTM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005677937.3 | XP_005677994.1 | 4 - 5 | 366 | 71 | |
| GenBank mRNA | JO356794.2 | - | 4 - 5 | 636 | 71 | |
| KA310044.1 | - | 4 - 5 | 383 | 71 | ||
More Information
Set Membership: |
|

GSTM3
Genomic Map