Assay Details
| Gene Symbol: LOC102191560 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005694976.3 | XP_005695033.2 | 2 - 3 | 239 | 102 | |
| GenBank mRNA | JO411441.2 | - | 2 - 3 | 233 | 102 | |
More Information
Set Membership: |
|

LOC102191560
Genomic Map