Assay Details
| Gene Symbol: KCNIP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018054834.1 | XP_017910323.1 | 5 - 6 | 570 | 65 | |
| GenBank mRNA | JO629486.2 | - | 5 - 6 | 513 | 65 | |
| KA346355.1 | - | 5 - 6 | 541 | 65 | ||
More Information
Set Membership: |
|

KCNIP3
Genomic Map