Assay Details
| Gene Symbol: EFHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018041119.1 | XP_017896608.1 | 2 - 3 | 504 | 68 | |
| GenBank mRNA | JO356366.2 | - | 2 - 3 | 355 | 68 | |
More Information
Set Membership: |
|

EFHD1
Genomic Map