Assay Details
| Gene Symbol: VRK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018055272.1 | XP_017910761.1 | 8 - 9 | 910 | 59 | |
| XM_018055273.1 | XP_017910762.1 | 8 - 9 | 931 | 59 | ||
| XM_018055274.1 | XP_017910763.1 | 7 - 8 | 558 | 59 | ||
| XM_018055275.1 | XP_017910764.1 | 8 - 9 | 916 | 59 | ||
More Information
Set Membership: |
|

VRK2
Genomic Map