Assay Details
| Gene Symbol: LOC102187635 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018058324.1 | XP_017913813.1 | 2 - 3 | 986 | 91 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102187635| Gene Symbol: LOC102187635 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018058324.1 | XP_017913813.1 | 2 - 3 | 986 | 91 | |
Set Membership: |
|