Assay Details
| Gene Symbol: THTPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005685236.3 | XP_005685293.2 | 2 - 3 | 912 | 94 | |
| XM_013966865.2 | XP_013822319.1 | 2 - 3 | 805 | 94 | ||
| GenBank mRNA | JO637915.2 | - | 2 - 3 | 94 | ||
| Gene Symbol: AP1G2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | KA311483.1 | - | 2 - 3 | 674 | 94 | |
More Information
Set Membership: |
|

THTPA
Genomic Map