Assay Details
| Gene Symbol: ALPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018055210.1 | XP_017910699.1 | 9 - 10 | 1267 | 57 | |
| XM_018055220.1 | XP_017910709.1 | 9 - 10 | 1225 | 57 | ||
| XM_018055232.1 | XP_017910721.1 | 10 - 11 | 1406 | 57 | ||
| XM_018055238.1 | XP_017910727.1 | 9 - 10 | 1238 | 57 | ||
| GenBank mRNA | JO623947.2 | - | 3 - 4 | 1265 | 57 | |
More Information
Set Membership: |
|

ALPL
Genomic Map