Assay Details
| Gene Symbol: BGN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018045199.1 | XP_017900688.1 | 5 - 6 | 837 | 66 | |
| GenBank mRNA | JO358343.2 | - | 3 - 4 | 1346 | 66 | |
| JO358344.2 | - | 3 - 4 | 1346 | 66 | ||
More Information
Set Membership: |
|

BGN
Genomic Map