Assay Details
| Gene Symbol: FMO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005690635.3 | XP_005690692.1 | 7 - 8 | 1305 | 62 | |
| GenBank mRNA | KA324842.1 | - | 7 - 8 | 1355 | 62 | |
More Information
Set Membership: |
|

FMO2
Genomic Map