Assay Details
| Gene Symbol: SNF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005693698.3 | XP_005693755.1 | 4 - 5 | 449 | 64 | |
| GenBank mRNA | JO512374.2 | - | 4 - 5 | 364 | 64 | |
| JO610654.2 | - | 4 - 5 | 558 | 64 | ||
More Information
Set Membership: |
|

SNF8
Genomic Map