Assay Details
| Gene Symbol: SIKE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005677832.3 | XP_005677889.2 | 3 - 4 | 502 | 77 | |
| GenBank mRNA | JT630152.1 | - | 3 - 4 | 362 | 77 | |
More Information
Set Membership: |
|

SIKE1
Genomic Map