Assay Details
| Gene Symbol: APIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018059250.1 | XP_017914739.1 | 2 - 3 | 297 | 72 | |
| GenBank mRNA | JO509236.2 | - | 5 - 6 | 629 | 72 | |
More Information
Set Membership: |
|

APIP
Genomic Map