Assay Details
| Gene Symbol: TSN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005676269.3 | XP_005676326.1 | 3 - 4 | 379 | 66 | |
| XM_005676270.3 | XP_005676327.1 | 3 - 4 | 378 | 66 | ||
| GenBank mRNA | JO576216.1 | - | 1539 | 66 | ||
More Information
Set Membership: |
|

TSN
Genomic Map