Assay Details
| Gene Symbol: YRDC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018041898.1 | XP_017897387.1 | 4 - 5 | 879 | 64 | |
| GenBank mRNA | KA350362.1 | - | 6 - 7 | 1682 | 64 | |
More Information
Set Membership: |
|

YRDC
Genomic Map