Assay Details
| Gene Symbol: KCTD15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018062282.1 | XP_017917771.1 | 5 - 6 | 667 | 58 | |
| XM_018062283.1 | XP_017917772.1 | 5 - 6 | 670 | 58 | ||
| XM_018062284.1 | XP_017917773.1 | 5 - 6 | 1637 | 58 | ||
| GenBank mRNA | JT678571.1 | - | 4 - 5 | 621 | 58 | |
More Information
Set Membership: |
|

KCTD15
Genomic Map