Assay Details
| Gene Symbol: PWWP3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018050448.1 | XP_017905937.1 | 8 - 9 | 1263 | 72 | |
| XM_018050449.1 | XP_017905938.1 | 8 - 9 | 1263 | 72 | ||
More Information
Set Membership: |
|

PWWP3A
Genomic Map