Assay Details
| Gene Symbol: RBM23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005685270.2 | XP_005685327.1 | 10 - 11 | 1177 | 57 | |
| XM_013966886.2 | XP_013822340.1 | 9 - 10 | 1818 | 57 | ||
| XM_018054567.1 | XP_017910056.1 | 10 - 11 | 1177 | 57 | ||
| XM_018054568.1 | XP_017910057.1 | 10 - 11 | 1049 | 57 | ||
| XM_018054569.1 | XP_017910058.1 | 13 - 14 | 1293 | 57 | ||
| XM_018054570.1 | XP_017910059.1 | 11 - 12 | 1240 | 57 | ||
More Information
Set Membership: |
|

RBM23
Genomic Map