Assay Details
| Gene Symbol: IRAK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005695610.2 | XP_005695667.2 | 11 - 12 | 1546 | 66 | |
| XM_005695611.3 | XP_005695668.2 | 11 - 12 | 1546 | 66 | ||
More Information
Set Membership: |
|

IRAK2
Genomic Map