Assay Details
| Gene Symbol: ELMOD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005691226.2 | XP_005691283.1 | 6 - 7 | 674 | 79 | |
| GenBank mRNA | JO417715.2 | - | 3 - 4 | 675 | 79 | |
More Information
Set Membership: |
|

ELMOD2
Genomic Map