Assay Details
| Gene Symbol: GAL3ST3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018042926.1 | XP_017898415.1 | 2 - 3 | 443 | 78 | |
| XM_018042927.1 | XP_017898416.1 | 2 - 3 | 404 | 78 | ||
More Information
Set Membership: |
|

GAL3ST3
Genomic Map