Assay Details
| Gene Symbol: SPOP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064907.1 | XP_017920396.1 | 5 - 6 | 622 | 64 | |
| XM_018064908.1 | XP_017920397.1 | 4 - 5 | 654 | 64 | ||
| XM_018064909.1 | XP_017920398.1 | 4 - 5 | 440 | 64 | ||
| GenBank mRNA | JO396369.2 | - | 6 - 7 | 872 | 64 | |
| JT676413.1 | - | 5 - 6 | 685 | 64 | ||
| JT747279.1 | - | 5 - 6 | 642 | 64 | ||
More Information
Set Membership: |
|

SPOP
Genomic Map