Assay Details
Gene Symbol: MINDY3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005687973.3 | XP_005688030.2 | 8 - 9 | 962 | 78 | |
XM_005687974.2 | XP_005688031.2 | 8 - 9 | 962 | 78 | ||
XM_018057150.1 | XP_017912639.1 | 7 - 8 | 909 | 78 | ||
GenBank mRNA | JT720458.1 | - | 9 - 10 | 1468 | 78 | |
KA330152.1 | - | 8 - 9 | 945 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
