Assay Details
| Gene Symbol: TRAPPC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005678700.3 | XP_005678757.1 | 2 - 3 | 272 | 100 | |
| XM_005678701.3 | XP_005678758.1 | 2 - 3 | 254 | 100 | ||
| XM_005678702.3 | XP_005678759.1 | 3 - 4 | 316 | 100 | ||
| XM_005678703.3 | XP_005678760.1 | 3 - 4 | 299 | 100 | ||
| XM_018041595.1 | XP_017897084.1 | 2 - 3 | 179 | 100 | ||
| GenBank mRNA | JO598585.2 | - | 3 - 4 | 1036 | 100 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005678702.3, XM_005678703.3, XM_018041595.1 transcripts.
Set Membership: |
|

TRAPPC3
Genomic Map