Assay Details
| Gene Symbol: CD180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005694612.2 | XP_005694669.1 | 2 - 3 | 381 | 102 | |
| GenBank mRNA | JO577905.1 | - | 2 - 3 | 465 | 102 | |
More Information
Set Membership: |
|

CD180
Genomic Map