Assay Details
| Gene Symbol: ABHD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039760.1 | XP_017895249.1 | 2 - 3 | 442 | 79 | |
| XM_018039761.1 | XP_017895250.1 | 2 - 3 | 442 | 79 | ||
| GenBank mRNA | JO643893.2 | - | 2 - 3 | 463 | 79 | |
More Information
Set Membership: |
|

ABHD3
Genomic Map