Assay Details
| Gene Symbol: ALG10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680680.3 | XP_005680737.1 | 1 - 2 | 473 | 70 | |
| XM_013964121.2 | XP_013819575.1 | 1 - 2 | 331 | 70 | ||
| GenBank mRNA | JO610019.2 | - | 1 - 2 | 309 | 70 | |
More Information
Set Membership: |
|

ALG10
Genomic Map