Assay Details
| Gene Symbol: PIP4K2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018048018.1 | XP_017903507.1 | 5 - 6 | 770 | 71 | |
| XM_018048019.1 | XP_017903508.1 | 5 - 6 | 683 | 71 | ||
More Information
Set Membership: |
|

PIP4K2C
Genomic Map