Assay Details
| Gene Symbol: NPTN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018054660.1 | XP_017910149.1 | 5 - 6 | 1032 | 61 | |
| XM_018054661.1 | XP_017910150.1 | 5 - 6 | 1032 | 61 | ||
| XM_018054663.1 | XP_017910152.1 | 4 - 5 | 684 | 61 | ||
| XM_018054664.1 | XP_017910153.1 | 4 - 5 | 684 | 61 | ||
| GenBank mRNA | JO632926.2 | - | 4 - 5 | 779 | 61 | |
More Information
Set Membership: |
|

NPTN
Genomic Map