Assay Details
| Gene Symbol: RRAGC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018042008.1 | XP_017897497.1 | 4 - 5 | 924 | 82 | |
| GenBank mRNA | JT693729.1 | - | 4 - 5 | 1021 | 82 | |
More Information
Set Membership: |
|

RRAGC
Genomic Map