Assay Details
| Gene Symbol: CD3EAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005692639.3 | XP_005692696.1 | 2 - 3 | 420 | 85 | |
| GenBank mRNA | JO616323.2 | - | 2 - 3 | 350 | 85 | |
More Information
Set Membership: |
|

CD3EAP
Genomic Map