Assay Details
| Gene Symbol: HMGA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039155.1 | XP_017894644.1 | 3 - 4 | 389 | 82 | |
| XM_018039156.1 | XP_017894645.1 | 3 - 4 | 392 | 82 | ||
More Information
Set Membership: |
|

HMGA1
Genomic Map