Assay Details
| Gene Symbol: TAGAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018053423.1 | XP_017908912.1 | 9 - 10 | 1065 | 71 | |
| GenBank mRNA | JO340288.2 | - | 1 - 2 | 439 | 71 | |
More Information
Set Membership: |
|

TAGAP
Genomic Map