Assay Details
| Gene Symbol: CPNE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005688504.3 | XP_005688561.1 | 4 - 5 | 435 | 89 | |
| XM_018057779.1 | XP_017913268.1 | 5 - 6 | 642 | 89 | ||
| XM_018057781.1 | XP_017913270.1 | 6 - 7 | 750 | 89 | ||
| XM_018057782.1 | XP_017913271.1 | 4 - 5 | 557 | 89 | ||
| XM_018057783.1 | XP_017913272.1 | 5 - 6 | 610 | 89 | ||
More Information
Set Membership: |
|

CPNE1
Genomic Map