Assay Details
| Gene Symbol: RAB7B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005690407.3 | XP_005690464.2 | 2 - 3 | 294 | 60 | |
| GenBank mRNA | KA349855.1 | - | 2 - 3 | 353 | 60 | |
More Information
Set Membership: |
|

RAB7B
Genomic Map