Assay Details
| Gene Symbol: SS18L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018057593.1 | XP_017913082.1 | 8 - 9 | 1179 | 54 | |
| GenBank mRNA | KA306864.1 | - | 4 - 5 | 523 | 54 | |
More Information
Set Membership: |
|

SS18L1
Genomic Map