Assay Details
| Gene Symbol: NRBF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018042540.1 | XP_017898029.1 | 3 - 4 | 366 | 66 | |
| XM_018042541.1 | XP_017898030.1 | 2 - 3 | 281 | 66 | ||
| GenBank mRNA | KA322293.1 | - | 3 - 4 | 333 | 66 | |
More Information
Set Membership: |
|

NRBF2
Genomic Map