Assay Details
| Gene Symbol: TRMT11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005684481.2 | XP_005684538.1 | 10 - 11 | 1050 | 75 | |
| XM_005684482.3 | XP_005684539.1 | 10 - 11 | 1050 | 75 | ||
| XM_013966364.2 | XP_013821818.1 | 10 - 11 | 1050 | 75 | ||
| GenBank mRNA | JO603025.2 | - | 5 - 6 | 505 | 75 | |
More Information
Set Membership: |
|

TRMT11
Genomic Map