Assay Details
| Gene Symbol: GFAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065253.1 | XP_017920742.1 | 9 - 10 | 1417 | 58 | |
| XM_018065254.1 | XP_017920743.1 | 8 - 9 | 1297 | 58 | ||
| GenBank mRNA | JO631891.2 | - | 6 - 7 | 703 | 58 | |
More Information
Set Membership: |
|

GFAP
Genomic Map