Assay Details
| Gene Symbol: LOC108633215 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018043671.1 | XP_017899160.1 | 11 - 12 | 1016 | 67 | |
| Gene Symbol: LOC102180115 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JO420746.2 | - | 1547 | 67 | ||
More Information
Set Membership: |
|

LOC108633215
Genomic Map