Assay Details
| Gene Symbol: LNX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005681612.3 | XP_005681669.2 | 4 - 5 | 1166 | 55 | |
| XM_005681613.3 | XP_005681670.2 | 4 - 5 | 1234 | 55 | ||
| XM_005681615.3 | XP_005681672.2 | 2 - 3 | 610 | 55 | ||
| XM_018049456.1 | XP_017904945.1 | 3 - 4 | 939 | 55 | ||
More Information
Set Membership: |
|

LNX1
Genomic Map