Assay Details
| Gene Symbol: LOC102188781 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005688013.3 | XP_005688070.1 | 2 - 3 | 227 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102188781| Gene Symbol: LOC102188781 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005688013.3 | XP_005688070.1 | 2 - 3 | 227 | 71 | |
Set Membership: |
|