Assay Details
| Gene Symbol: LOC102188781 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005688013.3 | XP_005688070.1 | 4 - 5 | 462 | 89 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102188781| Gene Symbol: LOC102188781 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005688013.3 | XP_005688070.1 | 4 - 5 | 462 | 89 | |
Set Membership: |
|